siRNA Details
| Gene Symbol: LINC01533 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110673.1 | 3 | 486 | |||
| NR_110674.1 | 3 | 330 | ||||
| GenBank mRNA | AK097493.1 | - | 3 | 486 | ||
| BX116841.1 | - | 3 | 296 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DB032536.1 | - | 3 | 486 | Chr. 19 - 32025862 - 32039595 [+] on Build GRCh38 | |

LINC01533
Genomic Map