siRNA Details
| Gene Symbol: LINC01533 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110673.1 | 2 | 246 | |||
| NR_110674.1 | 2 | 246 | ||||
| NR_110675.1 | 3 | 352 | ||||
| GenBank mRNA | AK097493.1 | - | 2 | 246 | ||
| BX116841.1 | - | 2 | 212 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01533| Gene Symbol: LINC01533 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110673.1 | 2 | 246 | |||
| NR_110674.1 | 2 | 246 | ||||
| NR_110675.1 | 3 | 352 | ||||
| GenBank mRNA | AK097493.1 | - | 2 | 246 | ||
| BX116841.1 | - | 2 | 212 | |||