siRNA Details
| Gene Symbol: LINC01533 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK097493.1 | - | 5 | 1491 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01533| Gene Symbol: LINC01533 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK097493.1 | - | 5 | 1491 | ||