siRNA Details
| Gene Symbol: MAGI2-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF521131.1 | - | 932 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MAGI2-IT1| Gene Symbol: MAGI2-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF521131.1 | - | 932 | |||