siRNA Details
| Gene Symbol: LOC283737 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK096897.1 | - | 1 | 1636 | ||
| Gene Symbol: AGBL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011521226.2 | XP_011519528.1 | 23 | 12488 | ||
| XM_017021918.1 | XP_016877407.1 | 23 | 12455 | |||
| XM_017021919.1 | XP_016877408.1 | 22 | 12404 | |||
| GenBank mRNA | AL133446.1 | - | 1 | 761 | ||

LOC283737
Genomic Map