siRNA Details
Gene Symbol: LINC00921 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033904.1 | 1 | 646 | |||
GenBank mRNA | BC104941.1 | - | 1 | 671 | ||
BC104943.1 | - | 1 | 671 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00921 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033904.1 | 1 | 646 | |||
GenBank mRNA | BC104941.1 | - | 1 | 671 | ||
BC104943.1 | - | 1 | 671 |