siRNA Details
| Gene Symbol: FFAR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC039436.1 | - | 1 | 457 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI603605.1 | - | 1 | 462 | Chr. 19 - 35352419 - 35353229 [+] on Build GRCh38 | |

FFAR1
Genomic Map