siRNA Details
Gene Symbol: CEP41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257160.1 | NP_001244089.1 | 3 | 3101 | ||
GenBank mRNA | AF429309.1 | - | 3 | 2947 | ||
AK001820.1 | - | 1 | 1846 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CEP41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257160.1 | NP_001244089.1 | 3 | 3101 | ||
GenBank mRNA | AF429309.1 | - | 3 | 2947 | ||
AK001820.1 | - | 1 | 1846 |