siRNA Details
| Gene Symbol: LOC101927793 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135194.1 | 3 | 2291 | |||
| GenBank mRNA | AK055866.1 | - | 3 | 2534 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927793| Gene Symbol: LOC101927793 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135194.1 | 3 | 2291 | |||
| GenBank mRNA | AK055866.1 | - | 3 | 2534 | ||