siRNA Details
| Gene Symbol: TTC14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001288582.1 | NP_001275511.1 | 13 | 2011 | ||
| GenBank mRNA | AY358762.1 | - | 13 | 1931 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BC065823.1 | - | 1 | 1596 | Chr. 3 - 180615889 - 180617620 [+] on Build GRCh38 | |

TTC14
Genomic Map