siRNA Details
Gene Symbol: APCDD1L-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK090612.1 | - | 1 | 251 | ||
AK091704.1 | - | 3 | 1596 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: APCDD1L-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK090612.1 | - | 1 | 251 | ||
AK091704.1 | - | 3 | 1596 |