siRNA Details
| Gene Symbol: PDCD6IP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BQ895877.1 | - | 1 | 409 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR622191.1 | - | 1 | 668 | Chr. 3 - 33797188 - 33799063 [-] on Build GRCh38 | |

PDCD6IP
Genomic Map