siRNA Details
| Gene Symbol: RBM41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001324242.1 | NP_001311171.1 | 8 | 2693 | ||
| NM_001324243.1 | NP_001311172.1 | 8 | 2786 | |||
| NM_018301.4 | NP_060771.3 | 7 | 2621 | |||
| NR_136728.1 | 7 | 2289 | ||||
| NR_136729.1 | 8 | 2383 | ||||
| NR_136730.1 | 8 | 3417 | ||||
| NR_136732.1 | 7 | 2382 | ||||
| XM_011530981.2 | XP_011529283.1 | 7 | 2713 | |||
| GenBank mRNA | AL832966.1 | - | 1 | 726 | ||

RBM41
Genomic Map