siRNA Details
| Gene Symbol: POLR2F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301130.1 | NP_001288059.1 | 6 | 1116 | ||
| NM_001301131.1 | NP_001288060.1 | 5 | 957 | |||
| XM_017028832.1 | XP_016884321.1 | 6 | 1004 | |||
| GenBank mRNA | AK026327.1 | - | 1 | 1303 | ||

POLR2F
Genomic Map