siRNA Details
| Gene Symbol: RASEF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152573.3 | NP_689786.2 | 17 | 2843 | ||
| XM_005251730.3 | XP_005251787.1 | 16 | 2823 | |||
| XM_005251731.3 | XP_005251788.1 | 17 | 3334 | |||
| GenBank mRNA | AK056882.1 | - | 1 | 119 | ||

RASEF
Genomic Map