siRNA Details
| Gene Symbol: LINC00930 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK093148.1 | - | 1195 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00930| Gene Symbol: LINC00930 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK093148.1 | - | 1195 | |||