siRNA Details
| Gene Symbol: DGAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AI126704.1 | - | 1 | 348 | ||
| Gene Symbol: LOC100506113 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL117542.1 | - | 3 | 435 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000534354.1 | - | 3 | 435 | Chr. 11 - 75800877 - 75803415 [-] on Build GRCh38 | |

DGAT2
Genomic Map