siRNA Details
| Gene Symbol: KCNMB2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | U61096.1 | - | 1 | 297 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KCNMB2-AS1| Gene Symbol: KCNMB2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | U61096.1 | - | 1 | 297 | ||