siRNA Details
| Gene Symbol: LOC107983947 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011510285.1 | XP_011508587.1 | 1 | 1058 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC107983947| Gene Symbol: LOC107983947 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011510285.1 | XP_011508587.1 | 1 | 1058 | ||