siRNA Details
| Gene Symbol: LOC107983947 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011510285.1 | XP_011508587.1 | 1 | 774 | ||
| GenBank mRNA | BC112311.1 | - | 1 | 816 | ||
| BC112313.1 | - | 1 | 816 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC107983947| Gene Symbol: LOC107983947 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011510285.1 | XP_011508587.1 | 1 | 774 | ||
| GenBank mRNA | BC112311.1 | - | 1 | 816 | ||
| BC112313.1 | - | 1 | 816 | |||