siRNA Details
| Gene Symbol: LOC157562 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL050061.1 | - | 1 | 1318 | ||
| BX649145.1 | - | 1 | 2091 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC157562| Gene Symbol: LOC157562 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL050061.1 | - | 1 | 1318 | ||
| BX649145.1 | - | 1 | 2091 | |||