siRNA Details
| Gene Symbol: CSNK1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC005905.1 | - | 2 | 467 | ||
| BC020745.1 | - | 2 | 452 | |||
| BX647478.1 | - | 2 | 3812 | |||
| BX648552.1 | - | 2 | 1341 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CSNK1A1| Gene Symbol: CSNK1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC005905.1 | - | 2 | 467 | ||
| BC020745.1 | - | 2 | 452 | |||
| BX647478.1 | - | 2 | 3812 | |||
| BX648552.1 | - | 2 | 1341 | |||