siRNA Details
| Gene Symbol: CSNK1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC020745.1 | - | 2 | 979 | ||
| BX648552.1 | - | 2 | 1868 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | L26953.1 | - | 796 | Chr. - - [-] on Build | ||

CSNK1A1
Genomic Map