siRNA Details
| Gene Symbol: ELP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011533851.2 | XP_011532153.1 | 5 | 645 | ||
| XM_011533852.2 | XP_011532154.1 | 5 | 981 | |||
| XM_011533853.2 | XP_011532155.1 | 5 | 622 | |||
| XM_011533854.2 | XP_011532156.1 | 4 | 546 | |||
| XM_011533856.2 | XP_011532158.1 | 4 | 601 | |||
| XM_011533857.2 | XP_011532159.1 | 4 | 771 | |||

ELP6
Genomic Map