siRNA Details
| Gene Symbol: LOC100288637 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038253.1 | 6 | 3192 | |||
| NR_038254.1 | 5 | 3092 | ||||
| NR_038255.1 | 4 | 2997 | ||||
| GenBank mRNA | AK092283.1 | - | 1 | 206 | ||
| AK095096.1 | - | 1 | 1117 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100288637| Gene Symbol: LOC100288637 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038253.1 | 6 | 3192 | |||
| NR_038254.1 | 5 | 3092 | ||||
| NR_038255.1 | 4 | 2997 | ||||
| GenBank mRNA | AK092283.1 | - | 1 | 206 | ||
| AK095096.1 | - | 1 | 1117 | |||