siRNA Details
| Gene Symbol: TCL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF195821.1 | - | 4 | 1160 | ||
| AK023714.1 | - | 1 | 1031 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TCL6| Gene Symbol: TCL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF195821.1 | - | 4 | 1160 | ||
| AK023714.1 | - | 1 | 1031 | |||