siRNA Details
| Gene Symbol: ZNF736 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170905.2 | NP_001164376.1 | 4 | 4929 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK125478.1 | - | 1 | 1280 | Chr. 7 - 64351235 - 64354685 [+] on Build GRCh38 | |

ZNF736
Genomic Map