siRNA Details
| Gene Symbol: ZNF254 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK092080.1 | - | 7 | 1091 | ||
| AK092150.1 | - | 4 | 765 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF254| Gene Symbol: ZNF254 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK092080.1 | - | 7 | 1091 | ||
| AK092150.1 | - | 4 | 765 | |||