siRNA Details
| Gene Symbol: BRCC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005274751.4 | XP_005274808.1 | 12 | 3428 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK098449.1 | - | 1 | 245 | Chr. X - 155123316 - 155124935 [+] on Build GRCh38 | |

BRCC3
Genomic Map