siRNA Details
| Gene Symbol: NKD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033119.4 | NP_149110.1 | 10 | 5721 | ||
| XM_011523399.2 | XP_011521701.1 | 13 | 6032 | |||
| XM_011523400.2 | XP_011521702.1 | 8 | 5323 | |||
| XM_017023804.1 | XP_016879293.1 | 12 | 7251 | |||
| GenBank mRNA | AK023784.1 | - | 1 | 273 | ||

NKD1
Genomic Map