siRNA Details
| Gene Symbol: NKD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033119.4 | NP_149110.1 | 10 | 6425 | ||
| XM_011523399.2 | XP_011521701.1 | 13 | 6736 | |||
| XM_011523400.2 | XP_011521702.1 | 8 | 6027 | |||
| XM_017023804.1 | XP_016879293.1 | 12 | 7955 | |||
| GenBank mRNA | AK021701.1 | - | 1 | 702 | ||
| AK023784.1 | - | 1 | 977 | |||

NKD1
Genomic Map