siRNA Details
Gene Symbol: LOC101928272 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120635.1 | 2 | 124 | |||
GenBank mRNA | AK127590.1 | - | 2 | 124 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA497137.1 | - | 2 | 124 | Chr. 10 - 9270282 - 9295593 [-] on Build GRCh38 |
