siRNA Details
| Gene Symbol: LOC101928272 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120635.1 | 5 | 1351 | |||
| GenBank mRNA | AK127590.1 | - | 5 | 1351 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928272| Gene Symbol: LOC101928272 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120635.1 | 5 | 1351 | |||
| GenBank mRNA | AK127590.1 | - | 5 | 1351 | ||