siRNA Details
| Gene Symbol: LINC00384 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX648491.1 | - | 1 | 2012 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000437721.1 | - | 1 | 2013 | Chr. 13 - 30151886 - 30159621 [-] on Build GRCh38 | |

LINC00384
Genomic Map