siRNA Details
Gene Symbol: LINC00260 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046213.1 | 1 | 945 | |||
GenBank mRNA | BC000988.2 | - | 1 | 945 | 3450898 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00260 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046213.1 | 1 | 945 | |||
GenBank mRNA | BC000988.2 | - | 1 | 945 | 3450898 |