siRNA Details
| Gene Symbol: LINC00260 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046213.1 | 1 | 494 | |||
| GenBank mRNA | BC000988.2 | - | 1 | 494 | 3450898 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00260| Gene Symbol: LINC00260 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046213.1 | 1 | 494 | |||
| GenBank mRNA | BC000988.2 | - | 1 | 494 | 3450898 | |