siRNA Details
| Gene Symbol: QKI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301085.1 | NP_001288014.1 | 8 | 6317 | ||
| NM_006775.2 | NP_006766.1 | 8 | 6341 | |||
| NM_206853.2 | NP_996735.1 | 7 | 11014 | |||
| NM_206854.2 | NP_996736.1 | 7 | 12293 | |||
| NM_206855.2 | NP_996737.1 | 6 | 13240 | |||
| XM_011536258.2 | XP_011534560.1 | 8 | 5845 | |||
| XM_011536260.2 | XP_011534562.1 | 7 | 6239 | |||
| GenBank mRNA | AK096701.1 | - | 1 | 1231 | ||

QKI
Genomic Map