siRNA Details
| Gene Symbol: LOC105375545 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_133932.1 | 1 | 180 | |||
| GenBank mRNA | DA339204.1 | - | 1 | 154 | ||
| DB173620.1 | - | 1 | 182 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105375545| Gene Symbol: LOC105375545 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_133932.1 | 1 | 180 | |||
| GenBank mRNA | DA339204.1 | - | 1 | 154 | ||
| DB173620.1 | - | 1 | 182 | |||