siRNA Details
| Gene Symbol: TMEM139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | CB241413.1 | - | 1 | 455 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AB074268.1 | - | 1 | 568 | Chr. 7 - 143285808 - 143288178 [-] on Build GRCh38 | |

TMEM139
Genomic Map