siRNA Details
| Gene Symbol: EHF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001206615.1 | NP_001193544.1 | 8 | 5354 | ||
| NM_001206616.1 | NP_001193545.1 | 9 | 5424 | |||
| NM_012153.5 | NP_036285.2 | 9 | 5423 | |||
| XM_005252857.1 | XP_005252914.1 | 9 | 5421 | |||
| XM_005252860.1 | XP_005252917.1 | 9 | 5277 | |||
| XM_005252861.1 | XP_005252918.1 | 8 | 5355 | |||
| XM_005252862.1 | XP_005252919.1 | 8 | 5224 | |||
| XM_011519983.1 | XP_011518285.1 | 9 | 5449 | |||
| XM_011519984.1 | XP_011518286.1 | 10 | 5529 | |||
| XM_011519985.1 | XP_011518287.1 | 9 | 5314 | |||
| XM_017017534.1 | XP_016873023.1 | 8 | 5352 | |||

EHF
Genomic Map