siRNA Details
| Gene Symbol: LOC101928591 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135065.1 | 1 | 49 | |||
| GenBank mRNA | AY726565.1 | - | 1 | 13 | ||
| BX109663.1 | - | 1 | 49 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928591| Gene Symbol: LOC101928591 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135065.1 | 1 | 49 | |||
| GenBank mRNA | AY726565.1 | - | 1 | 13 | ||
| BX109663.1 | - | 1 | 49 | |||