siRNA Details
| Gene Symbol: LOC101928591 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135065.1 | 9 | 1200 | |||
| GenBank mRNA | AY726565.1 | - | 9 | 1163 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928591| Gene Symbol: LOC101928591 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135065.1 | 9 | 1200 | |||
| GenBank mRNA | AY726565.1 | - | 9 | 1163 | ||