siRNA Details
| Gene Symbol: GDF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182828.3 | NP_878248.2 | 2 | 8841 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | U92981.1 | - | 1 | 529 | Chr. 2 - 20677809 - 20679237 [+] on Build GRCh38 | |

GDF7
Genomic Map