siRNA Details
| Gene Symbol: LINC01254 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110775.1 | 1 | 889 | |||
| GenBank mRNA | AY726567.1 | - | 1 | 889 | ||
| BC040323.1 | - | 1 | 1034 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01254| Gene Symbol: LINC01254 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110775.1 | 1 | 889 | |||
| GenBank mRNA | AY726567.1 | - | 1 | 889 | ||
| BC040323.1 | - | 1 | 1034 | |||