siRNA Details
| Gene Symbol: LSM11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173491.3 | NP_775762.1 | 4 | 2686 | ||
| GenBank mRNA | AK123951.1 | - | 1 | 940 | ||
| BC051353.1 | - | 4 | 2609 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK124248.1 | - | 1 | 1593 | Chr. 5 - 157755220 - 157890320 [+] on Build GRCh38 | |

LSM11
Genomic Map