siRNA Details
| Gene Symbol: GIMAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130759.3 | NP_570115.1 | 3 | 2607 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX356712.2 | - | 1 | 515 | Chr. 7 - 150721957 - 150723027 [+] on Build GRCh38 | |

GIMAP1
Genomic Map