siRNA Details
Gene Symbol: LOC101929341 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134310.1 | 2 | 781 | |||
GenBank mRNA | BC065517.1 | - | 2 | 781 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101929341 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134310.1 | 2 | 781 | |||
GenBank mRNA | BC065517.1 | - | 2 | 781 |