siRNA Details
| Gene Symbol: LOC101929341 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134310.1 | 1 | 295 | |||
| GenBank mRNA | BC065517.1 | - | 1 | 295 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BM554429.1 | - | 1 | 315 | Chr. 8 - 52194360 - 52199553 [+] on Build GRCh38 | |

LOC101929341
Genomic Map