siRNA Details
| Gene Symbol: CPOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX648974.1 | - | 1 | 127 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK022155.1 | - | 1 | 991 | Chr. 3 - 98582511 - 98584330 [-] on Build GRCh38 | |

CPOX
Genomic Map