siRNA Details
| Gene Symbol: LINC00299 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034135.1 | 2 | 298 | |||
| GenBank mRNA | AK127578.1 | - | 2 | 298 | ||
| BC043563.1 | - | 1 | 58 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00299| Gene Symbol: LINC00299 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034135.1 | 2 | 298 | |||
| GenBank mRNA | AK127578.1 | - | 2 | 298 | ||
| BC043563.1 | - | 1 | 58 | |||